4-Aminobutyl 4-hydroxy-3,5-dimethoxybenzoate hydrochloride 97%

Supplier: Ambeed
Warning

A1369242-100MG A1369242-1G A1369242-250MG
76659-504EA 132.62 USD
76659-504 76659-506 76659-508
4-Aminobutyl 4-hydroxy-3,5-dimethoxybenzoate hydrochloride 97%
4-Aminobutyl 4-hydroxy-3,5-dimethoxybenzoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₀ClNO₅
MW: 305.7546 g/mol
CAS Number: 18780-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR