2'-O-MOE-5-Me-C(Bz) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3R,4R,5R)-5-(4-Benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(2-methoxyethoxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
A1369224-5G
A1369224-1G
A1369224-25G
76659-502EA
400
USD
76659-502
77597-000
77867-633
2'-O-MOE-5-Me-C(Bz) 98%
2'-O-MOE-5-Me-C(Bz)
Precaución: for Research Use Only
Formula:
C₅₀H₆₀N₅O₁₀P MW: 922.01 g/mol |
CAS Number:
163759-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...