4-(Hydroxymethyl)-2-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A1369222-100MG A1369222-250MG
76659-498EA 111 USD
76659-498 76659-500
4-(Hydroxymethyl)-2-nitrobenzaldehyde 97%
4-(Hydroxymethyl)-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD29120359
CAS Number: 1289162-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR