3,6-Dimethyl-1,2,4,5-tetrazine 97%

Supplier: Ambeed
Warning

A1369174-250MG A1369174-100MG A1369174-1G
76659-496EA 183 USD
76659-496 77592-598 77596-994
3,6-Dimethyl-1,2,4,5-tetrazine 97%
3,6-Dimethyl-1,2,4,5-tetrazine

Precaución: for Research Use Only

Formula: C₄H₆N₄
MDL Number: MFCD09866053
CAS Number: 1558-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR