2-(4-Amino-2-nitrophenoxy)acetic acid 90%

Supplier: Ambeed
Warning

A1368330-100MG A1368330-250MG
76659-488EA 63.66 USD
76659-488 76659-490
2-(4-Amino-2-nitrophenoxy)acetic acid 90%
2-(4-Amino-2-nitrophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₈H₈N₂O₅
MW: 212.1595 g/mol
CAS Number: 1822951-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR