4-Amino-3-methyl-5-(prop-1-en-2-yl)benzonitrile 97%

Supplier: Ambeed
Warning

A1368278-1G A1368278-250MG A1368278-5G
76659-478EA 107.16 USD
76659-478 76659-480 76659-482
4-Amino-3-methyl-5-(prop-1-en-2-yl)benzonitrile 97%
4-Amino-3-methyl-5-(prop-1-en-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂
MW: 172.2264 g/mol
CAS Number: 2682115-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR