1,2-Bis(3-fluoro-4-methoxyphenyl)disulfane 97%

Supplier: Ambeed
Warning

A1368214-100MG A1368214-1G A1368214-250MG A1368214-5G
76659-464EA 25.47 USD
76659-464 76659-466 76659-468 76659-470
1,2-Bis(3-fluoro-4-methoxyphenyl)disulfane 97%
1,2-Bis(3-fluoro-4-methoxyphenyl)disulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂O₂S₂
MW: 314.3707 g/mol
CAS Number: 89818-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR