2,3,4,5-Tetrafluorophenol 97%

Supplier: Ambeed
Warning

A1368015-1G A1368015-250MG
76659-454EA 398.9 USD
76659-454 76659-456
2,3,4,5-Tetrafluorophenol 97%
2,3,4,5-Tetrafluorophenol

Precaución: for Research Use Only

Formula: C₆H₂F₄O
MW: 166.0731 g/mol
MDL Number: MFCD12032171
CAS Number: 3467-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR