[3-(Dimethylamino)propyl](propyl)amine 96%

Supplier: Ambeed
Danger

A1367927-1G A1367927-250MG A1367927-25G
76659-448EA 39 USD
76659-448 76659-450 77867-628
[3-(Dimethylamino)propyl](propyl)amine 96%
[3-(Dimethylamino)propyl](propyl)amine

Precaución: for Research Use Only

Formula: C₈H₂₀N₂
MDL Number: MFCD11146411
CAS Number: 19475-28-6
UN: 2734
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR