5-Methoxy-N,N,2-trimethylaniline 97%

Supplier: Ambeed
Warning

A1367884-250MG A1367884-5G
76659-444EA 70.02 USD
76659-444 76659-446
5-Methoxy-N,N,2-trimethylaniline 97%
5-Methoxy-N,N,2-trimethylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
MW: 165.2322 g/mol
MDL Number: MFCD25964628
CAS Number: 56140-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR