Methyl 4-(4-methylbenzoyl)-1H-pyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

A1367848-100MG A1367848-1G A1367848-250MG A1367848-5G
76659-436EA 21.22 USD
76659-436 76659-438 76659-440 76659-442
Methyl 4-(4-methylbenzoyl)-1H-pyrrole-2-carboxylate 97%
Methyl 4-(4-methylbenzoyl)-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₃
MW: 243.2579 g/mol
MDL Number: MFCD33022453
CAS Number: 2682115-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR