Boc-2-Cyclopropylglycine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-((tert-Butoxycarbonyl)amino)-2-cyclopropylacetic acid, Boc-DL-Cyclopropyl-Gly-OH
A1367162-100MG
A1367162-1G
A1367162-250MG
76659-404EA
35
USD
76659-404
76659-406
76659-408
Boc-2-Cyclopropylglycine 95%
Boc-2-Cyclopropylglycine
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₄ MW: 215.246 g/mol |
MDL Number:
MFCD01861823 CAS Number: 54256-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...