1,3-Bis(2,6-dibenzhydryl-4-methylphenyl)-1H-imidazol-3-ium chloride 95%
Supplier: Ambeed
|
Warning
|
A1366192-100MG
A1366192-1G
A1366192-250MG
76659-390EA
76.39
USD
76659-390
76659-392
76659-394
1,3-Bis(2,6-dibenzhydryl-4-methylphenyl)-1H-imidazol-3-ium chloride 95%
1,3-Bis(2,6-dibenzhydryl-4-methylphenyl)-1H-imidazol-3-ium chloride
Precaución: for Research Use Only
Formula:
C₆₉H₅₇ClN₂ MW: 949.6573 g/mol |
MDL Number:
MFCD31580175 CAS Number: 1218778-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...