4-(5,6-Dimethoxybenzo[b]thiophen-2-yl)-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: MSA-2

A1365634-100MG A1365634-1G A1365634-250MG A1365634-50MG A1365634-25MG
76659-380EA 551.67 USD
76659-380 76659-382 76659-384 76659-386 77596-974
4-(5,6-Dimethoxybenzo[b]thiophen-2-yl)-4-oxobutanoic acid 98%
4-(5,6-Dimethoxybenzo[b]thiophen-2-yl)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₅S
MW: 294.3230 g/mol
MDL Number: MFCD32640674
CAS Number: 129425-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR