2-(Isopropylamino)-3-methyl-5-(6-methyl-5-((2-(1-methyl-1H-pyrazol-4-yl)pyridin-4-yl)oxy)pyridin-2-yl)pyrimidin-4(3H)-one 98%

Supplier: Ambeed
Warning

Synonyms: Vimseltinib

A1365584-25MG A1365584-50MG
76659-376EA 182.48 USD
76659-376 76659-378
2-(Isopropylamino)-3-methyl-5-(6-methyl-5-((2-(1-methyl-1H-pyrazol-4-yl)pyridin-4-yl)oxy)pyridin-2-yl)pyrimidin-4(3H)-one 98%
2-(Isopropylamino)-3-methyl-5-(6-methyl-5-((2-(1-methyl-1H-pyrazol-4-yl)pyridin-4-yl)oxy)pyridin-2-yl)pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₂₃H₂₅N₇O₂
MW: 431.49 g/mol
MDL Number: MFCD32858282
CAS Number: 1628606-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR