(3,4-Difluoro-2-methylphenyl)methanamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A1364661-100MG
A1364661-250MG
76659-364EA
61.54
USD
76659-364
76659-366
(3,4-Difluoro-2-methylphenyl)methanamine hydrochloride 97%
(3,4-Difluoro-2-methylphenyl)methanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₀ClF₂N MW: 193.6215 g/mol |
CAS Number:
2703756-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...