Methyl 4-(1-cyanocyclopropyl)-3-fluorobenzoate 97%

Supplier: Ambeed
Warning

A1364611-100MG A1364611-1G A1364611-250MG A1364611-50MG
76659-352EA 23 USD
76659-352 76659-354 76659-356 76659-358
Methyl 4-(1-cyanocyclopropyl)-3-fluorobenzoate 97%
Methyl 4-(1-cyanocyclopropyl)-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₀FNO₂
MW: 219.2117 g/mol
MDL Number: MFCD19692682
CAS Number: 1314772-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR