6-(4-((4-Butylphenyl)diazenyl)phenoxy)hexyl methacrylate 98%

Supplier: Ambeed
Warning

A1364282-1G A1364282-250MG A1364282-5G
76659-336EA 101 USD
76659-336 76659-338 76659-340
6-(4-((4-Butylphenyl)diazenyl)phenoxy)hexyl methacrylate 98%
6-(4-((4-Butylphenyl)diazenyl)phenoxy)hexyl methacrylate

Precaución: for Research Use Only

Formula: C₂₆H₃₄N₂O₃
MW: 422.5598 g/mol
CAS Number: 1802983-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR