6-(4-(Phenyldiazenyl)phenoxy)hexyl methacrylate 98%

Supplier: Ambeed
Warning

A1364281-1G A1364281-250MG
76659-332EA 73 USD
76659-332 76659-334
6-(4-(Phenyldiazenyl)phenoxy)hexyl methacrylate 98%
6-(4-(Phenyldiazenyl)phenoxy)hexyl methacrylate

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₂O₃
MW: 366.4535 g/mol
CAS Number: 169237-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR