4,4'-(2,2-Diphenylethene-1,1-diyl)dibenzonitrile 98%

Supplier: Ambeed
Warning

A1364280-1G A1364280-250MG
76659-328EA 66.84 USD
76659-328 76659-330
4,4'-(2,2-Diphenylethene-1,1-diyl)dibenzonitrile 98%
4,4'-(2,2-Diphenylethene-1,1-diyl)dibenzonitrile

Precaución: for Research Use Only

Formula: C₂₈H₁₈N₂
MW: 382.4559 g/mol
CAS Number: 2323541-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR