Methyl 4-((4-bromophenyl)ethynyl)benzoate 98%

Supplier: Ambeed
Warning

A1364216-100MG A1364216-1G A1364216-250MG
76659-322EA 46.68 USD
76659-322 76659-324 76659-326
Methyl 4-((4-bromophenyl)ethynyl)benzoate 98%
Methyl 4-((4-bromophenyl)ethynyl)benzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₁BrO₂
MW: 315.1613 g/mol
MDL Number: MFCD28453504
CAS Number: 2365379-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR