2-(4'-(9H-Carbazol-9-yl)-[1,1'-biphenyl]-4-yl)acetonitrile 97%
Supplier: Ambeed
|
Warning
|
A1364114-1G
A1364114-250MG
76659-292EA
164.44
USD
76659-292
76659-294
2-(4'-(9H-Carbazol-9-yl)-[1,1'-biphenyl]-4-yl)acetonitrile 97%
2-(4'-(9H-Carbazol-9-yl)-[1,1'-biphenyl]-4-yl)acetonitrile
Precaución: for Research Use Only
Formula:
C₂₆H₁₈N₂ MW: 358.4345 g/mol |
CAS Number:
2370920-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...