1-(4-(1,2,2-Triphenylvinyl)phenyl)-1H-pyrrole-2,5-dione 97%

Supplier: Ambeed
Danger

A1364112-1G A1364112-250MG A1364112-5G
76659-286EA 122.01 USD
76659-286 76659-288 76659-290
1-(4-(1,2,2-Triphenylvinyl)phenyl)-1H-pyrrole-2,5-dione 97%
1-(4-(1,2,2-Triphenylvinyl)phenyl)-1H-pyrrole-2,5-dione

Precaución: for Research Use Only

Formula: C₃₀H₂₁NO₂
MW: 427.4933 g/mol
CAS Number: 2414485-55-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR