Methyl 4-carbamoyl-2-methoxybenzoate 97%

Supplier: Ambeed
Warning

A1364000-100MG A1364000-1G A1364000-250MG
76659-280EA 40 USD
76659-280 76659-282 76659-284
Methyl 4-carbamoyl-2-methoxybenzoate 97%
Methyl 4-carbamoyl-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD28403646
CAS Number: 1513849-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR