(S)-5-Amino-3-(4-((5-fluoro-2-methoxybenzamido)methyl)phenyl)-1-(1,1,1-trifluoropropan-2-yl)-1H-pyrazole-4-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: Pirtobrutinib

A1363918-100MG A1363918-250MG A1363918-50MG
76659-266EA 326.76 USD
76659-266 76659-268 76659-270
(S)-5-Amino-3-(4-((5-fluoro-2-methoxybenzamido)methyl)phenyl)-1-(1,1,1-trifluoropropan-2-yl)-1H-pyrazole-4-carboxamide 97%
(S)-5-Amino-3-(4-((5-fluoro-2-methoxybenzamido)methyl)phenyl)-1-(1,1,1-trifluoropropan-2-yl)-1H-pyrazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₂₂H₂₁F₄N₅O₃
MW: 479.43 g/mol
CAS Number: 2101700-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR