Methyl 2-chloro-6-cyanobenzoate 97%

Supplier: Ambeed
Warning

A1363890-100MG A1363890-250MG
76659-260EA 54.11 USD
76659-260 76659-262
Methyl 2-chloro-6-cyanobenzoate 97%
Methyl 2-chloro-6-cyanobenzoate

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MW: 195.6024 g/mol
MDL Number: MFCD18398897
CAS Number: 58331-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR