3,4,5-Trimethoxy-N-(4-(8-methylimidazo[1,2-a]pyridin-2-yl)phenyl)benzamide 97%

Supplier: Ambeed
Warning

A1363652-100MG A1363652-1G A1363652-250MG
76659-238EA 85.94 USD
76659-238 76659-240 76659-242
3,4,5-Trimethoxy-N-(4-(8-methylimidazo[1,2-a]pyridin-2-yl)phenyl)benzamide 97%
3,4,5-Trimethoxy-N-(4-(8-methylimidazo[1,2-a]pyridin-2-yl)phenyl)benzamide

Precaución: for Research Use Only

Formula: C₂₄H₂₃N₃O₄
MW: 417.4571 g/mol
MDL Number: MFCD04092556
CAS Number: 724737-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR