Methyl 4-((5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoate 98%

Supplier: Ambeed
Warning

A1363348-1G A1363348-250MG
76659-224EA 109.28 USD
76659-224 76659-226
Methyl 4-((5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoate 98%
Methyl 4-((5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoate

Precaución: for Research Use Only

Formula: C₂₃H₂₇NO₃
MW: 365.4654 g/mol
MDL Number: MFCD12828201
CAS Number: 94497-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR