3,6,11,14-Tetrabromodibenzo[a,c]triphenylene 98%

Supplier: Ambeed
Warning

A1363255-100MG A1363255-1G A1363255-250MG
76659-214EA 50.93 USD
76659-214 76659-216 76659-218
3,6,11,14-Tetrabromodibenzo[a,c]triphenylene 98%
3,6,11,14-Tetrabromodibenzo[a,c]triphenylene

Precaución: for Research Use Only

Formula: C₂₆H₁₂Br₄
MW: 643.9895 g/mol
MDL Number: MFCD33022443
CAS Number: 2135768-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR