(R)-4-Benzyl-2-(2-chloro-6-(diphenylphosphino)phenyl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1363155-100MG A1363155-1G A1363155-250MG
76659-196EA 27 USD
76659-196 76659-198 76659-200
(R)-4-Benzyl-2-(2-chloro-6-(diphenylphosphino)phenyl)-4,5-dihydrooxazole ≥97%, ee 99%
(R)-4-Benzyl-2-(2-chloro-6-(diphenylphosphino)phenyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₈H₂₃ClNOP
CAS Number: 2634687-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR