(2-((2-Methoxyethoxy)methyl)phenyl)methanamine 97%

Supplier: Ambeed
Danger

A1363063-100MG A1363063-1G A1363063-250MG
76659-188EA 27 USD
76659-188 76659-190 76659-192
(2-((2-Methoxyethoxy)methyl)phenyl)methanamine 97%
(2-((2-Methoxyethoxy)methyl)phenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₂
MDL Number: MFCD09807682
CAS Number: 1016512-39-2
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR