tert-Butyl 4-(pyrimidin-2-yl)benzylcarbamate 97%

Supplier: Ambeed
Warning

A1363049-100MG A1363049-1G A1363049-250MG
76659-182EA 192.03 USD
76659-182 76659-184 76659-186
tert-Butyl 4-(pyrimidin-2-yl)benzylcarbamate 97%
tert-Butyl 4-(pyrimidin-2-yl)benzylcarbamate

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₃O₂
MW: 285.3410 g/mol
MDL Number: MFCD33022437
CAS Number: 2682114-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR