5-(4-([1,1'-Biphenyl]-4-yl)quinazolin-2-yl)-8-phenyl-5,8-dihydroindolo[2,3-c]carbazole 97%

Supplier: Ambeed
Warning

A1362915-1G A1362915-250MG A1362915-5G
76659-168EA 102.91 USD
76659-168 76659-170 76659-172
5-(4-([1,1'-Biphenyl]-4-yl)quinazolin-2-yl)-8-phenyl-5,8-dihydroindolo[2,3-c]carbazole 97%
5-(4-([1,1'-Biphenyl]-4-yl)quinazolin-2-yl)-8-phenyl-5,8-dihydroindolo[2,3-c]carbazole

Precaución: for Research Use Only

Formula: C₄₄H₂₈N₄
MW: 612.7199 g/mol
CAS Number: 2159132-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR