(2-((tert-Butoxycarbonyl)amino)-7-fluorobenzo[d]thiazol-4-yl)boronic acid 97%

Supplier: Ambeed
Warning

A1362869-100MG A1362869-1G A1362869-250MG
76659-162EA 92 USD
76659-162 76659-164 76659-166
(2-((tert-Butoxycarbonyl)amino)-7-fluorobenzo[d]thiazol-4-yl)boronic acid 97%
(2-((tert-Butoxycarbonyl)amino)-7-fluorobenzo[d]thiazol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄BFN₂O₄S
MDL Number: MFCD32689945
CAS Number: 2415163-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR