(R)-4-(1-Aminoethyl)benzenesulfonamide hydrochloride 97%

Supplier: Ambeed
Warning

A1362671-250MG A1362671-1G
76659-160EA 342 USD
76659-160 77566-708
(R)-4-(1-Aminoethyl)benzenesulfonamide hydrochloride 97%
(R)-4-(1-Aminoethyl)benzenesulfonamide hydrochloride

Precaución: For research use only.

Formula: C₈H₁₃ClN₂O₂S
MW: 236.7190 g/mol
MDL Number: MFCD32205395
CAS Number: 1203655-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR