2'-Bromospiro[adamantane-2,9'-fluorene] 97%

Supplier: Ambeed
Warning

A1362587-1G A1362587-250MG A1362587-5G
76659-144EA 45 USD
76659-144 76659-146 77867-549
2'-Bromospiro[adamantane-2,9'-fluorene] 97%
2'-Bromospiro[adamantane-2,9'-fluorene]

Precaución: for Research Use Only

Formula: C₂₂H₂₁Br
CAS Number: 2407620-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR