4-(p-Tolyl)-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indole-7-carbaldehyde 97%

Supplier: Ambeed
Warning

A1362527-1G A1362527-250MG A1362527-5G
76659-138EA 143 USD
76659-138 76659-140 76659-142
4-(p-Tolyl)-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indole-7-carbaldehyde 97%
4-(p-Tolyl)-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indole-7-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO
MW: 277.3603 g/mol
CAS Number: 273220-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR