(R)-2-(6-(5-Chloro-2-((tetrahydro-2H-pyran-4-yl)amino)pyrimidin-4-yl)-1-oxoisoindolin-2-yl)-N-((S)-1-(3-fluoro-5-methoxyphenyl)-2-hydroxyethyl)propanamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
ASTX029
A1362224-1MG
A1362224-25MG
A1362224-5MG
76659-124EA
48.81
USD
76659-124
76659-126
76659-128
(R)-2-(6-(5-Chloro-2-((tetrahydro-2H-pyran-4-yl)amino)pyrimidin-4-yl)-1-oxoisoindolin-2-yl)-N-((S)-1-(3-fluoro-5-methoxyphenyl)-2-hydroxyethyl)propanamide 98%
(R)-2-(6-(5-Chloro-2-((tetrahydro-2H-pyran-4-yl)amino)pyrimidin-4-yl)-1-oxoisoindolin-2-yl)-N-((S)-1-(3-fluoro-5-methoxyphenyl)-2-hydroxyethyl)propanamide
Precaución: for Research Use Only
Formula:
C₂₉H₃₁ClFN₅O₅ MW: 584.04 g/mol |
MDL Number:
MFCD32874105 CAS Number: 2095719-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...