(4-((2-(2,5-Dimethylphenoxy)ethyl)amino)piperidin-1-yl)(pyridin-4-yl)methanone 98%

Supplier: Ambeed
Warning

Synonyms: GRP-60367

A1362216-10MG A1362216-1MG A1362216-25MG A1362216-50MG A1362216-5MG
76659-114EA 60.48 USD
76659-114 76659-116 76659-118 76659-120 76659-122
(4-((2-(2,5-Dimethylphenoxy)ethyl)amino)piperidin-1-yl)(pyridin-4-yl)methanone 98%
(4-((2-(2,5-Dimethylphenoxy)ethyl)amino)piperidin-1-yl)(pyridin-4-yl)methanone

Precaución: for Research Use Only

Formula: C₂₁H₂₇N₃O₂
MW: 353.4580 g/mol
MDL Number: MFCD19043296
CAS Number: 1309241-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR