(E)-2-([1,1'-Biphenyl]-4-ylamino)-N-hydroxy-2-oxoacetimidoyl cyanide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
DHODH-IN-11
A1361930-10MG
A1361930-1MG
A1361930-25MG
A1361930-50MG
A1361930-5MG
76659-104EA
50.93
USD
76659-104
76659-106
76659-108
76659-110
76659-112
(E)-2-([1,1'-Biphenyl]-4-ylamino)-N-hydroxy-2-oxoacetimidoyl cyanide 98%
(E)-2-([1,1'-Biphenyl]-4-ylamino)-N-hydroxy-2-oxoacetimidoyl cyanide
Precaución: for Research Use Only
Formula:
C₁₅H₁₁N₃O₂ MW: 265.2667 g/mol |
MDL Number:
MFCD32663119 CAS Number: 1263303-95-2 UN: 1325 ADR: 4.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...