tert-Butyl 6-iodohexanoate 98+% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilized with Copper chip

A1361707-250MG A1361707-5G A1361707-100MG A1361707-1G
76659-100EA 66 USD
76659-100 76659-102 77596-884 77621-756
tert-Butyl 6-iodohexanoate 98+% stabilized
tert-Butyl 6-iodohexanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₉IO₂
MDL Number: MFCD27979336
CAS Number: 67899-04-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR