1,1'-Oxybis(propan-2-ol) 97% (mixture of isomers)

Supplier: Ambeed
Warning

A1361241-1000G A1361241-100G A1361241-500G
76659-092EA 87 USD
76659-092 76659-094 76659-096
1,1'-Oxybis(propan-2-ol) 97% (mixture of isomers)
1,1'-Oxybis(propan-2-ol)

Precaución: for Research Use Only

Formula: C₆H₁₄O₃
MDL Number: MFCD00004538
CAS Number: 110-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR