6-Amino-3,3-dimethylindolin-2-one 95%

Supplier: Ambeed
Warning

A1361217-1G A1361217-250MG A1361217-5G
76659-086EA 148.53 USD
76659-086 76659-088 76659-090
6-Amino-3,3-dimethylindolin-2-one 95%
6-Amino-3,3-dimethylindolin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.21 g/mol
MDL Number: MFCD11053509
CAS Number: 100510-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR