(S)-Ethyl 2,3-bis((tert-butoxycarbonyl)amino)propanoate 97%

Supplier: Ambeed
Warning

A1361097-1G A1361097-250MG A1361097-5G
76659-080EA 103.97 USD
76659-080 76659-082 76659-084
(S)-Ethyl 2,3-bis((tert-butoxycarbonyl)amino)propanoate 97%
(S)-Ethyl 2,3-bis((tert-butoxycarbonyl)amino)propanoate

Precaución: for Research Use Only

Formula: C₁₅H₂₈N₂O₆
MW: 332.3926 g/mol
MDL Number: MFCD22418850
CAS Number: 860033-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR