m-Cresol purple sodium salt indicator

Supplier: Ambeed
Warning

Synonyms: m-Cresolsulphonphthalein sodium salt, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[d][1,2]oxathiole 2,2-dioxide, sodium salt

A1360945-100G A1360945-1G A1360945-25G A1360945-5G
76659-058EA 275 USD
76659-058 76659-060 76659-062 76659-064
m-Cresol purple sodium salt indicator
m-Cresol purple sodium salt

Precaución: for Research Use Only

Formula: C₂₁H₁₇NaO₅S
MDL Number: MFCD00010177
CAS Number: 62625-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR