N-(1-(2,4-Bis(trifluoromethyl)benzyl)-1H-pyrazol-4-yl)-5-(furan-2-yl)isoxazole-3-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: Ceapin-A7

A1360823-10MG A1360823-1MG A1360823-25MG A1360823-50MG A1360823-5MG
76659-042EA 203.7 USD
76659-042 76659-044 76659-046 76659-048 76659-050
N-(1-(2,4-Bis(trifluoromethyl)benzyl)-1H-pyrazol-4-yl)-5-(furan-2-yl)isoxazole-3-carboxamide 97%
N-(1-(2,4-Bis(trifluoromethyl)benzyl)-1H-pyrazol-4-yl)-5-(furan-2-yl)isoxazole-3-carboxamide

Precaución: for Research Use Only

Formula: C₂₀H₁₂F₆N₄O₃
MW: 470.3247 g/mol
MDL Number: MFCD31567506
CAS Number: 2323027-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR