(2S,4S)-4-(Benzyloxy)-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A1360652-100MG
A1360652-250MG
76659-032EA
81.69
USD
76659-032
76659-034
(2S,4S)-4-(Benzyloxy)-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid 98%
(2S,4S)-4-(Benzyloxy)-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₇H₂₃NO₅ MW: 321.3682 g/mol |
MDL Number:
MFCD02094395 CAS Number: 54631-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...