3-(2,2,2-Trifluoroethoxy)pyridin-4-amine hydrochloride 95%

Supplier: Ambeed
Warning

A1360507-100MG A1360507-250MG
76659-028EA 116.7 USD
76659-028 76659-030
3-(2,2,2-Trifluoroethoxy)pyridin-4-amine hydrochloride 95%
3-(2,2,2-Trifluoroethoxy)pyridin-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClF₃N₂O
MW: 228.5994 g/mol
MDL Number: MFCD33022420
CAS Number: 2682112-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR