2,3-Dihydrofuro[2,3-b]pyridin-6-amine 97%

Supplier: Ambeed
Warning

A1360475-100MG A1360475-1G A1360475-250MG
76659-020EA 153.84 USD
76659-020 76659-022 76659-024
2,3-Dihydrofuro[2,3-b]pyridin-6-amine 97%
2,3-Dihydrofuro[2,3-b]pyridin-6-amine

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.1512 g/mol
MDL Number: MFCD28635568
CAS Number: 1550730-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR