4,6-Dibromo-3-fluoro-2-methoxyaniline 97%

Supplier: Ambeed
Warning

A1360421-100MG A1360421-250MG
76659-016EA 31.83 USD
76659-016 76659-018
4,6-Dibromo-3-fluoro-2-methoxyaniline 97%
4,6-Dibromo-3-fluoro-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₆Br₂FNO
MW: 298.9350 g/mol
MDL Number: MFCD33022418
CAS Number: 2682115-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR